Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCKAP1L SIRNA Assays ›
          Gene Symbol:
          Nckap1l
          Gene Name:
          NCK associated protein 1 like
          Gene Aliases:
          4930568P13Rik, AI463083, Hem1, Hemp1
          Chromosome Location:
          Chr.15: 103453784 - 103502860 on Build GRCm38
          UniGene ID:
          Mm.30805
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS200633
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nckap1l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153505.4 NP_705725.1 13 1309
          GenBank mRNA AK040192.1 - 13 1294
          AK083390.1 - 13 1312
          AK134039.1 - 13 1289
          AK149862.1 - 13 1308
          BC007186.1 - 13 1188
          BC037096.1 - 13 1303 5344383

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          myeloid cell homeostasis
          protein complex assembly
          chemotaxis
          maintenance of cell polarity
          neutrophil chemotaxis
          positive regulation of actin filament polymerization
          cortical actin cytoskeleton organization
          positive regulation of B cell proliferation
          negative regulation of interleukin-17 production
          negative regulation of interleukin-6 production
          positive regulation of cell adhesion mediated by integrin
          negative regulation of myosin-light-chain-phosphatase activity
          positive regulation of T cell proliferation
          positive regulation of phosphorylation
          response to drug
          T cell homeostasis
          negative regulation of apoptotic process
          positive regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of CD8-positive, alpha-beta T cell differentiation
          positive regulation of GTPase activity
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of lymphocyte differentiation
          positive regulation of erythrocyte differentiation
          positive regulation of protein kinase activity
          B cell receptor signaling pathway
          positive regulation of phagocytosis, engulfment
          actin polymerization-dependent cell motility
          positive regulation of neutrophil chemotaxis
          GTPase activator activity
          protein kinase activator activity
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0