Search
Search

Baz2a| Gene Symbol: Baz2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054078.2 | NP_473419.2 | 26 | 5121 | ||
| GenBank mRNA | AJ309544.2 | - | 24 | 4855 | ||
| AK084735.1 | - | 2 | 303 | |||
| AK122243.1 | - | 9 | 1967 | |||
| AK135732.1 | - | 6 | 1131 | |||
| AK140558.1 | - | 5 | 938 | |||
| AK149353.1 | - | 3 | 495 | |||
| BC078632.1 | - | 6 | 1582 | |||