Search
Search

Arg1| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007482.3 | NP_031508.1 | 8 | 1025 | ||
| GenBank mRNA | AK149471.1 | - | 8 | 1008 | ||
| AK171417.1 | - | 7 | 859 | |||
| BC013341.1 | - | 8 | 1002 | 4163626 | ||
| BC050005.1 | - | 8 | 1039 | 3990246 | ||
| U51805.1 | - | 8 | 1064 | |||