Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL3 SIRNA Assays ›
Gene Symbol:
Bcl3
Gene Name:
B cell leukemia/lymphoma 3
Gene Aliases:
AI528691, Bcl-3
Chromosome Location:
Chr.7: 19808462 - 19822811 on Build GRCm38
UniGene ID:
Mm.439658
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS202354
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Bcl3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033601.3
NP_291079.2
6
921
GenBank mRNA
AF067774.1
-
6
882
AK155016.1
-
6
886
BC021610.1
-
2
78
M90397.1
-
6
836
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
follicular dendritic cell differentiation
marginal zone B cell differentiation
humoral immune response mediated by circulating immunoglobulin
germinal center formation
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
I-kappaB kinase/NF-kappaB signaling
response to virus
response to UV-C
antimicrobial humoral response
extracellular matrix organization
DNA damage response, signal transduction by p53 class mediator
positive regulation of interferon-gamma production
T-helper 1 type immune response
negative regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
defense response to protozoan
regulation of apoptotic process
negative regulation of apoptotic process
T-helper 2 cell differentiation
positive regulation of interleukin-10 biosynthetic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of translation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
spleen development
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0