Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP8 SIRNA Assays ›
          Gene Symbol:
          Casp8 Gm20257
          Gene Name:
          caspase 8
          caspase 8 pseudogene
          Gene Aliases:
          CASP-8, FLICE, MACH, Mch5
          -
          Chromosome Location:
          Chr.1: 58795229 - 58847503 on Build GRCm38
          Chromosome Location:
          Chr.1: 58652641 - 58655831 on Build GRCm38
          UniGene ID:
          Mm.336851
          Mm.440933
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID MSS202641
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Casp8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080126.1 NP_001073595.1 9 1569
          NM_001277926.1 NP_001264855.1 10 1685
          NM_009812.2 NP_033942.1 9 1742
          GenBank mRNA AF067834.1 - 9 1622
          AJ000641.1 - 8 1188
          AJ007749.1 - 9 1395
          AK146758.1 - 9 1742
          AK153340.1 - 9 1451
          AK156135.1 - 9 1458
          AK168812.1 - 9 1742
          BC006737.1 - 9 1684 3964119
          BC049955.1 - 9 1576 4208824
          CT010166.1 - 8 1349
          Gene Symbol: Gm20257
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK079804.1 - 2 88

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          neural tube formation
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          heart development
          extrinsic apoptotic signaling pathway via death domain receptors
          macrophage differentiation
          response to tumor necrosis factor
          TRAIL-activated apoptotic signaling pathway
          regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of proteolysis
          cardiac muscle tissue development
          protein heterooligomerization
          proteolysis involved in cellular protein catabolic process
          negative regulation of necroptotic process
          regulation of thymocyte apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          execution phase of apoptosis
          activation of cysteine-type endopeptidase activity
          hepatocyte apoptotic process
          regulation of apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway
          endopeptidase activity
          cysteine-type endopeptidase activity
          death receptor binding
          tumor necrosis factor receptor binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          ubiquitin protein ligase binding
          protein complex binding
          death effector domain binding
          protein heterodimerization activity
          scaffold protein binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0