Search
Search

Cd19| Gene Symbol: Cd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009844.2 | NP_033974.2 | 2 | 278 | ||
| GenBank mRNA | AK089835.1 | - | 2 | 277 | ||
| AK156125.1 | - | 2 | 281 | |||
| AK163608.1 | - | 1 | 500 | |||
| AK172008.1 | - | 2 | 240 | |||
| BY234629.1 | - | 2 | 299 | |||