Search
Search

Col6a1| Gene Symbol: Col6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009933.4 | NP_034063.1 | 35 | 2795 | ||
| GenBank mRNA | BC002194.1 | - | 6 | 808 | ||
| BM453821.1 | - | 4 | 550 | |||
| X66405.1 | - | 35 | 2795 | |||
| Z18271.1 | - | 16 | 1341 | |||