Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSF1R SIRNA Assays ›
          Gene Symbol:
          Csf1r
          Gene Name:
          colony stimulating factor 1 receptor
          Gene Aliases:
          AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSF-R, M-CSFR
          Chromosome Location:
          Chr.18: 61096808 - 61131139 on Build GRCm38
          UniGene ID:
          Mm.22574
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS203291
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Csf1r
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037859.2 NP_001032948.2 4 861
          GenBank mRNA AK004947.1 - 3 671
          AK041299.1 - 3 649
          AK076215.1 - 3 669
          AK079247.1 - 3 678
          AK080326.1 - 3 658
          AK089498.1 - 3 651
          AK143545.1 - 3 662
          AK146069.1 - 4 862
          AK154128.1 - 3 676
          AK154556.1 - 3 672
          AK154653.1 - 3 672
          AK154699.1 - 3 672
          AK155488.1 - 3 681
          AK155565.1 - 3 691
          AK155576.1 - 678
          AK155640.1 - 3 676
          AK155654.1 - 3 676
          AK155858.1 - 3 679
          AK156550.1 - 3 696
          AK171241.1 - 3 711
          AK171386.1 - 3 676
          AK171543.1 - 3 651
          BC036343.1 - 3 648 4948135
          BC043054.1 - 3 652 6418718
          BC050024.1 - 3 718
          BC066863.1 - 4 770 30436119
          X06368.1 - 3 654
          X68932.1 - 3 654

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: X61479.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          immune system process
          protein phosphorylation
          inflammatory response
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell proliferation
          positive regulation of cell proliferation
          regulation of cell shape
          phosphorylation
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          hemopoiesis
          osteoclast differentiation
          positive regulation of cell migration
          ruffle organization
          cellular response to macrophage colony-stimulating factor stimulus
          macrophage colony-stimulating factor signaling pathway
          positive regulation of tyrosine phosphorylation of Stat3 protein
          innate immune response
          regulation of bone resorption
          cell-cell junction maintenance
          positive regulation of osteoclast differentiation
          phosphatidylinositol metabolic process
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of protein tyrosine kinase activity
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of chemokine secretion
          positive regulation of cell motility
          regulation of actin cytoskeleton reorganization
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          macrophage colony-stimulating factor receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein phosphatase binding
          cytokine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0