Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSN2 SIRNA Assays ›
Gene Symbol:
Csn2
Gene Name:
casein beta
Gene Aliases:
Csnb
Chromosome Location:
Chr.5: 87692619 - 87699429 on Build GRCm38
UniGene ID:
Mm.268737
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS203302
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Csn2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286020.1
NP_001272949.1
7
387
NM_001286021.1
NP_001272950.1
7
402
NM_001286022.1
NP_001272951.1
6
363
NM_001286023.1
NP_001272952.1
5
345
NM_001286024.1
NP_001272953.1
5
342
NM_009972.2
NP_034102.1
7
390
GenBank mRNA
AK021324.1
-
7
384
AK021328.1
-
7
384
AK052803.1
-
7
387
AK052805.1
-
7
387
AK052822.1
-
7
384
AK085678.1
-
7
384
AK085694.1
-
7
387
AK085698.1
-
7
384
AK085729.1
-
7
387
AK085756.1
-
7
384
AK142616.1
-
7
387
AK142620.1
-
7
387
AK142631.1
-
7
387
AK142635.1
-
7
387
AK164747.1
-
7
387
AK164754.1
-
2
1737
AK164761.1
-
7
387
AK164791.1
-
7
387
AK164792.1
-
7
384
AK164793.1
-
7
384
AK164794.1
-
7
387
BC003863.1
-
2
212
BC003976.1
-
3
236
BC013332.1
-
7
387
3670912
BC019114.1
-
7
368
3483918
BC019189.1
-
6
380
5028194
BC020356.1
-
7
367
BC020383.1
-
7
367
BC021153.1
-
7
385
3670372
BC050270.1
-
7
367
BC057671.1
-
4
2179
BC080709.1
-
7
371
3671502
X04490.1
-
7
386
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: J00711.
Panther Classification:
Molecular Function -
storage protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
calcium ion transport
lactation
negative regulation of catalytic activity
negative regulation of lactation
enzyme inhibitor activity
transporter activity
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0