Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HCN1 SIRNA Assays ›
Gene Symbol:
Hcn1
Gene Name:
hyperpolarization-activated, cyclic nucleotide-gated K+ 1
Gene Aliases:
Bcng1, C630013B14Rik, HAC2
Chromosome Location:
Chr.13: 117602320 - 117987414 on Build GRCm38
UniGene ID:
Mm.343429
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS205063
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hcn1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010408.3
NP_034538.2
2
1079
GenBank mRNA
AF028737.1
-
2
699
AJ225123.1
-
2
919
AK158322.1
-
2
1080
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
transport
ion transport
potassium ion transport
sodium ion transport
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
apical protein localization
negative regulation of action potential
retinal cone cell development
response to calcium ion
transmembrane transport
cellular response to cAMP
potassium ion transmembrane transport
regulation of membrane hyperpolarization
positive regulation of cation channel activity
nucleotide binding
ion channel activity
intracellular cAMP activated cation channel activity
voltage-gated ion channel activity
voltage-gated sodium channel activity
voltage-gated potassium channel activity
potassium channel activity
sodium channel activity
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
protein C-terminus binding
cAMP binding
protein complex binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0