Search
Search

Hnrnpab| Gene Symbol: Hnrnpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048061.1 | NP_001041526.1 | 3 | 543 | ||
| NM_010448.3 | NP_034578.1 | 3 | 543 | |||
| GenBank mRNA | AK013384.1 | - | 3 | 510 | ||
| AK013709.1 | - | 509 | ||||
| AK076132.1 | - | 3 | 506 | |||
| AK165609.1 | - | 4 | 490 | |||
| BC043069.1 | - | 3 | 497 | 6402930 | ||
| CA325334.1 | - | 2 | 241 | |||
| D90151.1 | - | 3 | 498 | |||
| DQ397821.1 | - | 2 | 305 | |||
| DQ397822.1 | - | 2 | 305 | |||
| L36663.1 | - | 502 | ||||