Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ICAM1 SIRNA Assays ›
          Gene Symbol:
          Icam1
          Gene Name:
          intercellular adhesion molecule 1
          Gene Aliases:
          CD54, Icam-1, Ly-47, MALA-2
          Chromosome Location:
          Chr.9: 21015960 - 21028797 on Build GRCm38
          UniGene ID:
          Mm.435508
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS205331
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Icam1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010493.2 NP_034623.1 5 1095
          GenBank mRNA AK149748.1 - 5 1071
          AK149781.1 - 5 1070
          AK149945.1 - 5 1070
          AK150003.1 - 5 1071
          AK150049.1 - 5 1070
          AK150057.1 - 5 1071
          AK151227.1 - 5 1071
          AK151681.1 - 5 1071
          AK152155.1 - 5 1071
          AK152527.1 - 5 1071
          AK152530.1 - 5 1071
          AK152556.1 - 5 1071
          AK156417.1 - 5 1071
          AK168275.1 - 5 1071
          AK171520.1 - 4 808
          AK172321.1 - 5 1071
          BC008626.1 - 5 1050 3588949
          CT010246.1 - 5 1020
          CT010302.1 - 5 1020
          M31585.1 - 5 1078
          X16624.1 - 5 1049
          X52264.1 - 5 1042

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          acute inflammatory response to antigenic stimulus
          T cell antigen processing and presentation
          positive regulation of cellular extravasation
          cell adhesion
          leukocyte cell-cell adhesion
          sensory perception of sound
          regulation of cell shape
          single organismal cell-cell adhesion
          regulation of cell adhesion
          positive regulation of actin filament polymerization
          cell adhesion mediated by integrin
          positive regulation of GTPase activity
          adhesion of symbiont to host
          positive regulation of nitric oxide biosynthetic process
          positive regulation of vasoconstriction
          receptor-mediated virion attachment to host cell
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of calcium ion transport
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to glucose stimulus
          establishment of Sertoli cell barrier
          regulation of ruffle assembly
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          integrin binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0