Search
Search

Itpr2| Gene Symbol: Itpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010586.2 | NP_034716.1 | 29 | 4078 | ||
| NM_019923.4 | NP_064307.2 | 30 | 4177 | |||
| GenBank mRNA | AB182288.1 | - | 30 | 3866 | ||
| AB182290.1 | - | 29 | 3767 | |||
| GU980658.1 | - | 30 | 3832 | |||