Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIF SIRNA Assays ›
          Gene Symbol:
          Lif
          Gene Name:
          leukemia inhibitory factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 4257568 - 4272514 on Build GRCm38
          UniGene ID:
          Mm.4964
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS205969
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lif
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039537.2 NP_001034626.1 3 532
          NM_008501.2 NP_032527.1 4 450
          GenBank mRNA AF065917.1 - 3 242
          AF065918.1 - 3 242
          AK042018.1 - 3 234
          AK155959.1 - 3 292
          BY209921.1 - 3 292
          X06381.1 - 2 179
          X12810.1 - 3 242

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          transcription from RNA polymerase II promoter
          immune response
          embryo implantation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of neuron projection development
          negative regulation of angiogenesis
          stem cell population maintenance
          lung development
          organ regeneration
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of MAPK cascade
          regulation of cell differentiation
          positive regulation of macrophage differentiation
          negative regulation of meiotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          negative regulation of hormone secretion
          lung alveolus development
          muscle organ morphogenesis
          neuron development
          astrocyte differentiation
          positive regulation of astrocyte differentiation
          leukemia inhibitory factor signaling pathway
          stem cell differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of corticotropin secretion
          retina development in camera-type eye
          maternal process involved in female pregnancy
          transdifferentiation
          lung vasculature development
          lung lobe morphogenesis
          trophoblast giant cell differentiation
          spongiotrophoblast differentiation
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of protein localization to nucleus
          positive regulation of histone H3-K27 acetylation
          regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor recruiting
          receptor binding
          cytokine activity
          leukemia inhibitory factor receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0