Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NODAL SIRNA Assays ›
          Gene Symbol:
          Nodal
          Gene Name:
          nodal
          Gene Aliases:
          Tg.413d
          Chromosome Location:
          Chr.10: 61417972 - 61425337 on Build GRCm38
          UniGene ID:
          Mm.57195
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS207086
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nodal
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013611.4 NP_038639.2 2 916
          GenBank mRNA AK049290.1 - 2 600
          AK082823.1 - 2 600
          BC128018.1 - 2 593 40136148
          X70514.1 - 2 915

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          gastrulation with mouth forming second
          mesoderm formation
          trophectodermal cell differentiation
          trophectodermal cellular morphogenesis
          neural fold formation
          liver development
          placenta development
          embryonic placenta development
          maternal placenta development
          vasculature development
          heart looping
          inhibition of neuroepithelial cell differentiation
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          determination of left/right symmetry
          gastrulation
          nervous system development
          brain development
          endoderm development
          heart development
          positive regulation of cell proliferation
          embryonic pattern specification
          anterior/posterior axis specification
          anterior/posterior pattern specification
          regulation of signal transduction
          polarity specification of proximal/distal axis
          regulation of gastrulation
          positive regulation of vascular endothelial growth factor production
          negative regulation of cell development
          positive regulation of pathway-restricted SMAD protein phosphorylation
          cell migration
          stem cell population maintenance
          positive regulation of cell-cell adhesion
          lung development
          positive regulation of activin receptor signaling pathway
          floor plate morphogenesis
          embryonic heart tube development
          endodermal cell differentiation
          nodal signaling pathway
          growth
          cell migration involved in gastrulation
          regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of MAPK cascade
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          axial mesodermal cell fate specification
          mesendoderm development
          cell development
          digestive tract morphogenesis
          anatomical structure formation involved in morphogenesis
          embryonic cranial skeleton morphogenesis
          tissue morphogenesis
          formation of anatomical boundary
          positive regulation of epithelial cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          digestive system development
          embryonic process involved in female pregnancy
          maternal process involved in parturition
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          left lung morphogenesis
          negative regulation of androgen receptor signaling pathway
          epiblast cell-extraembryonic ectoderm cell signaling involved in anterior/posterior axis specification
          positive regulation of ERK1 and ERK2 cascade
          primitive streak formation
          transforming growth factor beta receptor signaling pathway involved in primitive streak formation
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          negative regulation of trophoblast cell migration
          negative regulation of chorionic trophoblast cell proliferation
          positive regulation of receptor activity
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          receptor agonist activity
          type I activin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0