Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OXTR SIRNA Assays ›
Gene Symbol:
Oxtr
Gene Name:
oxytocin receptor
Gene Aliases:
OTR
Chromosome Location:
Chr.6: 112473684 - 112491308 on Build GRCm38
UniGene ID:
Mm.333310
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS207347
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Oxtr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081147.1
NP_001074616.1
1
790
GenBank mRNA
AK143340.1
-
1
755
D86599.1
-
3
1343
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012871, XM_008763182, XM_008763181, XM_006237008, XM_008763180, and others.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
suckling behavior
response to amphetamine
regulation of systemic arterial blood pressure by vasopressin
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
female pregnancy
lactation
memory
positive regulation of norepinephrine secretion
sleep
positive regulation of synaptic transmission, GABAergic
cellular response to hormone stimulus
response to anoxia
social behavior
maternal behavior
sperm ejaculation
eating behavior
regulation of digestive system process
positive regulation of blood pressure
positive regulation of vasoconstriction
response to steroid hormone
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
positive regulation of penile erection
negative regulation of gastric acid secretion
ERK1 and ERK2 cascade
positive regulation of uterine smooth muscle contraction
response to peptide
signal transducer activity
G-protein coupled receptor activity
oxytocin receptor activity
vasopressin receptor activity
peptide hormone binding
peptide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0