Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAX2 SIRNA Assays ›
Gene Symbol:
Pax2
Gene Name:
paired box 2
Gene Aliases:
Opdc, Pax-2
Chromosome Location:
Chr.19: 44745695 - 44837871 on Build GRCm38
UniGene ID:
Mm.192158
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS207405
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pax2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011037.4
NP_035167.4
7
838
GenBank mRNA
BC148232.1
-
7
842
40142573
BC150484.1
-
7
842
40142579
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106361, XM_008760403, XM_006231443, XM_008760405, XM_008760404.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
urogenital system development
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
cell fate determination
kidney development
mesonephros development
neural tube closure
optic cup morphogenesis involved in camera-type eye development
retinal pigment epithelium development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
mesodermal cell fate specification
positive regulation of cell proliferation
glial cell differentiation
optic nerve development
optic nerve morphogenesis
optic nerve structural organization
vestibulocochlear nerve formation
cell differentiation
regulation of metanephros size
ureter maturation
inner ear morphogenesis
regulation of apoptotic process
camera-type eye development
negative regulation of apoptotic process
regulation of programmed cell death
negative regulation of programmed cell death
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of cytolysis
positive regulation of transcription from RNA polymerase II promoter
animal organ development
pronephros development
brain morphogenesis
stem cell differentiation
positive regulation of epithelial cell proliferation
oxidation-reduction process
mesenchymal to epithelial transition
paramesonephric duct development
optic chiasma development
cellular response to glucose stimulus
renal system development
metanephric mesenchyme development
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
metanephric mesenchymal cell differentiation
mesonephric tubule development
mesonephric tubule formation
mesonephric duct development
nephric duct formation
ureter development
metanephric collecting duct development
metanephric distal convoluted tubule development
metanephric nephron tubule formation
positive regulation of metanephric glomerulus development
negative regulation of mesenchymal cell apoptotic process involved in metanephric nephron morphogenesis
regulation of metanephric nephron tubule epithelial cell differentiation
reactive oxygen species metabolic process
cochlea development
cochlea morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of mesenchymal cell apoptotic process involved in metanephros development
negative regulation of apoptotic process involved in metanephric collecting duct development
negative regulation of apoptotic process involved in metanephric nephron tubule development
negative regulation of reactive oxygen species metabolic process
positive regulation of metanephric DCT cell differentiation
positive regulation of optic nerve formation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
core promoter proximal region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
superoxide-generating NADPH oxidase activity
transcription regulatory region DNA binding
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0