Search
Search

Per1| Gene Symbol: Per1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159367.1 | NP_001152839.1 | 11 | 1537 | ||
| NM_011065.4 | NP_035195.2 | 11 | 1552 | |||
| GenBank mRNA | AB002108.1 | - | 10 | 1366 | ||
| AF022992.1 | - | 11 | 1591 | |||
| AK081813.1 | - | 11 | 1550 | |||
| AK148202.1 | - | 11 | 1544 | |||
| AK154900.1 | - | 11 | 1533 | |||
| AK172958.1 | - | 11 | 1537 | |||
| BC039768.1 | - | 10 | 1437 | 4163009 | ||
| BC091645.1 | - | 11 | 1539 | 30355259 | ||