Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3CD SIRNA Assays ›
          Gene Symbol:
          Pik3cd
          Gene Name:
          phosphatidylinositol 3-kinase catalytic delta polypeptide
          Gene Aliases:
          2410099E07Rik, 2610208K16Rik, AW545373, p110delta
          Chromosome Location:
          Chr.4: 149649168 - 149702566 on Build GRCm38
          UniGene ID:
          Mm.229108
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS207621
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pik3cd
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001029837.2 NP_001025008.2 10 1541
          NM_001164049.1 NP_001157521.1 10 1568
          NM_001164050.1 NP_001157522.1 10 1568
          NM_001164051.1 NP_001157523.1 10 1592
          NM_001164052.1 NP_001157524.1 8 1295
          NM_008840.3 NP_032866.2 11 1743
          GenBank mRNA AK040867.1 - 10 1503
          AK149935.1 - 11 1642
          AK150649.1 - 10 1469
          AK151482.1 - 10 1469
          AK152661.1 - 10 1469
          AK153304.1 - 10 1491
          AK155835.1 - 10 1454
          AK171025.1 - 10 1551
          AK172222.1 - 10 1445
          BC035203.1 - 10 1558 4192906
          CT010349.1 - 8 1263
          U86587.1 - 8 1263

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108978.

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          adaptive immune response
          immune system process
          protein phosphorylation
          chemotaxis
          inflammatory response
          cell surface receptor signaling pathway
          positive regulation of gene expression
          phosphatidylinositol 3-kinase signaling
          phosphorylation
          cell differentiation
          platelet activation
          positive regulation of cell migration
          phosphatidylinositol-3-phosphate biosynthetic process
          B cell activation
          innate immune response
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          homeostasis of number of cells
          defense response to fungus
          cell chemotaxis
          nucleotide binding
          protein binding
          ATP binding
          kinase activity
          1-phosphatidylinositol-3-kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0