Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLA1 SIRNA Assays ›
Gene Symbol:
Pola1
Gene Name:
polymerase (DNA directed), alpha 1
Gene Aliases:
AW321876, Pola
Chromosome Location:
Chr.X: 93304766 - 93632202 on Build GRCm38
UniGene ID:
Mm.1923
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS207822
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pola1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008892.2
NP_032918.1
20
2091
GenBank mRNA
D13543.1
-
20
2068
D17384.1
-
20
2091
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_016937, X06745.
This siRNA also targets the following transcripts in Rat: NM_053479, AJ011605.
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
DNA-dependent DNA replication
DNA replication, synthesis of RNA primer
DNA replication initiation
DNA strand elongation involved in DNA replication
leading strand elongation
lagging strand elongation
double-strand break repair via nonhomologous end joining
cell proliferation
DNA biosynthetic process
nucleotide binding
nucleoside binding
nucleic acid binding
DNA binding
chromatin binding
double-stranded DNA binding
DNA-directed DNA polymerase activity
DNA primase activity
protein binding
3'-5' exonuclease activity
transferase activity
nucleotidyltransferase activity
purine nucleotide binding
pyrimidine nucleotide binding
protein kinase binding
metal ion binding
protein heterodimerization activity
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0