Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS208037
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
9
1067
NM_001146121.1
NP_001139593.1
10
1073
NM_001146122.1
NP_001139594.1
9
1064
NM_001146123.1
NP_001139595.1
9
1040
NM_001146124.1
NP_001139596.1
9
983
NM_011179.3
NP_035309.3
10
1076
GenBank mRNA
AK088369.1
-
9
981
AK089998.1
-
9
985
AK093881.1
-
9
981
AK135637.1
-
9
981
AK145607.1
-
9
988
AK145802.1
-
9
982
AK148152.1
-
9
1008
AK148545.1
-
10
992
AK149785.1
-
9
984
AK150331.1
-
9
983
AK150645.1
-
9
981
AK151034.1
-
9
981
AK151173.1
-
9
982
AK151230.1
-
9
981
AK151234.1
-
9
956
AK151251.1
-
9
954
AK151525.1
-
9
983
AK151529.1
-
9
956
AK151686.1
-
9
1017
AK151852.1
-
9
983
AK151875.1
-
9
980
AK151891.1
-
9
981
AK151907.1
-
9
983
AK151943.1
-
9
983
AK152117.1
-
9
983
AK152126.1
-
9
981
AK152134.1
-
9
983
AK152203.1
-
9
1015
AK152280.1
-
9
981
AK152319.1
-
9
985
AK152355.1
-
9
983
AK152408.1
-
9
986
AK152505.1
-
9
983
AK152557.1
-
9
983
AK152605.1
-
9
983
AK152819.1
-
9
995
AK153404.1
-
9
981
AK159108.1
-
9
1011
AK159164.1
-
9
981
AK159167.1
-
9
982
AK159198.1
-
9
981
AK159221.1
-
9
982
AK159244.1
-
9
978
AK159266.1
-
9
981
AK159333.1
-
9
981
AK159341.1
-
9
981
AK159384.1
-
9
981
AK159414.1
-
9
984
AK159427.1
-
9
983
AK159451.1
-
9
981
AK159457.1
-
9
995
AK159463.1
-
9
983
AK159487.1
-
9
981
AK159492.1
-
9
978
AK159510.1
-
9
981
AK159528.1
-
9
981
AK159530.1
-
9
981
AK159589.1
-
9
983
AK159604.1
-
9
981
AK159622.1
-
8
873
AK159634.1
-
9
981
AK159638.1
-
9
982
AK159657.1
-
9
981
AK159664.1
-
9
981
AK159685.1
-
9
981
AK159692.1
-
9
981
AK159709.1
-
9
981
AK159720.1
-
9
981
AK159722.1
-
9
981
AK159728.1
-
9
983
AK159840.1
-
9
981
AK159861.1
-
9
981
AK159862.1
-
9
983
AK159877.1
-
9
981
AK159910.1
-
9
981
AK159915.1
-
9
981
AK159936.1
-
9
983
AK159982.1
-
9
981
AK159994.1
-
9
981
AK160015.1
-
9
981
AK160019.1
-
9
983
AK160021.1
-
9
981
AK160024.1
-
9
981
AK160046.1
-
9
981
AK160049.1
-
9
983
AK160073.1
-
9
981
AK160108.1
-
9
981
AK160877.1
-
9
984
AK161313.1
-
9
998
AK166700.1
-
9
981
AK166707.1
-
9
980
AK166744.1
-
9
979
AK166811.1
-
9
981
AK166849.1
-
9
981
AK166879.1
-
9
983
AK166909.1
-
9
981
AK166920.1
-
9
981
AK166946.1
-
9
986
AK166959.1
-
9
981
AK166984.1
-
9
981
AK167014.1
-
9
981
AK167025.1
-
9
981
AK167073.1
-
9
981
AK167099.1
-
9
984
AK167108.1
-
9
981
AK167111.1
-
9
981
AK167147.1
-
9
981
AK167198.1
-
9
988
AK167213.1
-
9
981
AK167237.1
-
9
981
AK167243.1
-
9
982
AK167252.1
-
9
983
AK167418.1
-
9
983
AK167719.1
-
9
983
AK167902.1
-
9
1051
AK170416.1
-
9
980
AK182197.1
-
3
218
AK191456.1
-
3
224
AK195069.1
-
3
224
AK196371.1
-
3
224
BC036266.1
-
8
847
BC141089.1
-
10
986
9055701
BC145359.1
-
9
980
9053917
S36200.1
-
10
966
U27340.1
-
9
981
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline