Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSAP SIRNA Assays ›
          Gene Symbol:
          Psap
          Gene Name:
          prosaposin
          Gene Aliases:
          AI037048, SGP-1
          Chromosome Location:
          Chr.10: 60277628 - 60302600 on Build GRCm38
          UniGene ID:
          Mm.277498
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS208038
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Psap
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146120.1 NP_001139592.1 2, 3 297
          NM_001146121.1 NP_001139593.1 2, 3 297
          NM_001146122.1 NP_001139594.1 2, 3 297
          NM_001146123.1 NP_001139595.1 2, 3 297
          NM_001146124.1 NP_001139596.1 2, 3 213
          NM_011179.3 NP_035309.3 2, 3 297
          GenBank mRNA AK088369.1 - 2, 3 211
          AK089998.1 - 2, 3 215
          AK093881.1 - 2, 3 211
          AK135637.1 - 2, 3 211
          AK145607.1 - 2, 3 213
          AK145802.1 - 2, 3 212
          AK148150.1 - 2, 3 219
          AK148152.1 - 2, 3 238
          AK148545.1 - 2, 3 213
          AK149785.1 - 2, 3 214
          AK150331.1 - 2, 3 213
          AK150645.1 - 2, 3 211
          AK151034.1 - 2, 3 211
          AK151173.1 - 2, 3 212
          AK151230.1 - 2, 3 211
          AK151234.1 - 2, 3 213
          AK151251.1 - 2, 3 211
          AK151525.1 - 2, 3 213
          AK151529.1 - 2, 3 213
          AK151686.1 - 2, 3 247
          AK151852.1 - 2, 3 213
          AK151875.1 - 2, 3 211
          AK151891.1 - 2, 3 211
          AK151907.1 - 2, 3 213
          AK151943.1 - 2, 3 213
          AK152117.1 - 2, 3 213
          AK152126.1 - 2, 3 211
          AK152134.1 - 2, 3 213
          AK152203.1 - 2, 3 245
          AK152280.1 - 2, 3 211
          AK152319.1 - 2, 3 215
          AK152355.1 - 2, 3 213
          AK152408.1 - 2, 3 216
          AK152505.1 - 2, 3 213
          AK152557.1 - 2, 3 213
          AK152605.1 - 2, 3 213
          AK152819.1 - 2, 3 225
          AK153404.1 - 2, 3 211
          AK159108.1 - 2, 3 241
          AK159164.1 - 2, 3 211
          AK159167.1 - 2, 3 212
          AK159198.1 - 2, 3 211
          AK159221.1 - 2, 3 215
          AK159244.1 - 2, 3 211
          AK159266.1 - 2, 3 211
          AK159333.1 - 2, 3 211
          AK159341.1 - 2, 3 211
          AK159384.1 - 2, 3 211
          AK159414.1 - 2, 3 214
          AK159427.1 - 2, 3 213
          AK159451.1 - 2, 3 211
          AK159457.1 - 2, 3 225
          AK159463.1 - 2, 3 213
          AK159487.1 - 2, 3 211
          AK159492.1 - 2, 3 211
          AK159510.1 - 2, 3 211
          AK159528.1 - 2, 3 211
          AK159530.1 - 2, 3 211
          AK159589.1 - 2, 3 213
          AK159604.1 - 2, 3 211
          AK159622.1 - 1, 2 103
          AK159634.1 - 2, 3 211
          AK159638.1 - 2, 3 212
          AK159657.1 - 2, 3 211
          AK159664.1 - 2, 3 211
          AK159685.1 - 2, 3 211
          AK159692.1 - 2, 3 211
          AK159709.1 - 2, 3 211
          AK159720.1 - 2, 3 211
          AK159722.1 - 2, 3 211
          AK159728.1 - 2, 3 213
          AK159840.1 - 2, 3 211
          AK159861.1 - 2, 3 211
          AK159862.1 - 2, 3 213
          AK159877.1 - 2, 3 211
          AK159910.1 - 2, 3 211
          AK159915.1 - 2, 3 211
          AK159936.1 - 2, 3 213
          AK159982.1 - 2, 3 211
          AK159994.1 - 2, 3 211
          AK160015.1 - 2, 3 211
          AK160019.1 - 2, 3 213
          AK160021.1 - 2, 3 211
          AK160024.1 - 2, 3 211
          AK160046.1 - 2, 3 211
          AK160049.1 - 2, 3 213
          AK160073.1 - 2, 3 211
          AK160108.1 - 2, 3 211
          AK160877.1 - 2, 3 214
          AK161313.1 - 2, 3 228
          AK166700.1 - 2, 3 211
          AK166707.1 - 2, 3 210
          AK166744.1 - 2, 3 209
          AK166811.1 - 2, 3 211
          AK166849.1 - 2, 3 211
          AK166879.1 - 2, 3 213
          AK166909.1 - 2, 3 211
          AK166920.1 - 2, 3 211
          AK166946.1 - 2, 3 211
          AK166959.1 - 2, 3 211
          AK166984.1 - 2, 3 211
          AK167014.1 - 2, 3 211
          AK167025.1 - 2, 3 211
          AK167073.1 - 2, 3 211
          AK167099.1 - 2, 3 214
          AK167108.1 - 2, 3 211
          AK167111.1 - 2, 3 211
          AK167147.1 - 2, 3 211
          AK167198.1 - 2, 3 213
          AK167213.1 - 2, 3 211
          AK167237.1 - 2, 3 211
          AK167243.1 - 2, 3 212
          AK167252.1 - 2, 3 213
          AK167418.1 - 2, 3 213
          AK167719.1 - 2, 3 213
          AK167902.1 - 2, 3 281
          AK170416.1 - 2, 3 210
          AK180024.1 - 2, 3 218
          AK180106.1 - 1, 2 67
          AK194829.1 - 1, 2 67
          AK197804.1 - 1, 2 67
          AK201095.1 - 1, 2 67
          AK210181.1 - 1, 2 81
          AK217017.1 - 2, 3 218
          BC036266.1 - 1, 2 77
          BC141089.1 - 2, 3 210 9055701
          BC145359.1 - 2, 3 210 9053917
          BY325400.1 - 2, 3 297
          S36200.1 - 2, 3 187
          S71616.1 - 2, 3 230
          U27340.1 - 2, 3 211

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          sphingolipid metabolic process
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          regulation of lipid metabolic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          developmental growth
          prostate gland growth
          epithelial cell differentiation involved in prostate gland development
          cellular response to organic substance
          negative regulation of hydrogen peroxide-induced cell death
          G-protein coupled receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline