Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLIT2 SIRNA Assays ›
          Gene Symbol:
          Slit2
          Gene Name:
          slit homolog 2 (Drosophila)
          Gene Aliases:
          Drad-1, E030015M03Rik, E130320P19Rik, Slil3, mKIAA4141, slit-2
          Chromosome Location:
          Chr.5: 47983108 - 48306282 on Build GRCm38
          UniGene ID:
          Mm.289739
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS209186
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slit2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291227.1 NP_001278156.1 33, 34 5483
          NM_001291228.1 NP_001278157.1 31, 32 5432
          NM_178804.4 NP_848919.3 31, 32 5420
          GenBank mRNA AF074960.1 - 17, 18 2047
          AF144628.1 - 31, 32 3781
          AK144573.1 - 31, 32 5378
          AK220505.1 - 31, 32 4016
          BC059267.1 - 15, 16 1809
          BC079911.1 - 31, 32 3545
          BC145487.1 - 33, 34 3827 9054050
          BC145488.1 - 31, 32 3776 9054051
          BC150779.1 - 33, 34 3827 9087690

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_022632, XM_008770210.

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          ureteric bud development
          in utero embryonic development
          negative regulation of protein phosphorylation
          cell migration involved in sprouting angiogenesis
          negative regulation of leukocyte chemotaxis
          chemotaxis
          multicellular organism development
          nervous system development
          axonogenesis
          axon guidance
          motor neuron axon guidance
          negative regulation of cell proliferation
          negative regulation of lamellipodium assembly
          negative regulation of endothelial cell migration
          negative regulation of gene expression
          negative regulation of smooth muscle cell migration
          single organismal cell-cell adhesion
          olfactory bulb development
          chemorepulsion involved in postnatal olfactory bulb interneuron migration
          corticospinal neuron axon guidance through spinal cord
          telencephalon cell migration
          cell differentiation
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of axon extension
          negative regulation of actin filament polymerization
          retinal ganglion cell axon guidance
          dorsal/ventral axon guidance
          negative regulation of GTPase activity
          Roundabout signaling pathway
          positive regulation of apoptotic process
          negative regulation of vascular permeability
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          axon extension involved in axon guidance
          negative regulation of inflammatory response
          negative chemotaxis
          induction of negative chemotaxis
          negative regulation of small GTPase mediated signal transduction
          mammary gland duct morphogenesis
          mammary duct terminal end bud growth
          negative regulation of chemokine-mediated signaling pathway
          cellular response to heparin
          negative regulation of smooth muscle cell chemotaxis
          negative regulation of mononuclear cell migration
          negative regulation of neutrophil chemotaxis
          negative regulation of monocyte chemotaxis
          negative regulation of retinal ganglion cell axon guidance
          negative regulation of cellular response to growth factor stimulus
          GTPase inhibitor activity
          receptor binding
          calcium ion binding
          protein binding
          heparin binding
          identical protein binding
          protein homodimerization activity
          laminin-1 binding
          proteoglycan binding
          heparan sulfate proteoglycan binding
          chemorepellent activity
          Roundabout binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline