Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNS2 SIRNA Assays ›
          Gene Symbol:
          Tns2
          Gene Name:
          tensin 2
          Gene Aliases:
          C1-ten, Tenc1, nep, nph
          Chromosome Location:
          Chr.15: 102100367 - 102116401 on Build GRCm38
          UniGene ID:
          Mm.29389
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS209762
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tns2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153533.2 NP_705761.2 18 2527
          GenBank mRNA AF424789.1 - 18 2397
          AK147789.1 - 18 2582
          AK147941.1 - 18 2586
          AK158009.1 - 18 2496
          BC025818.1 - 1 174
          BC036265.1 - 17 2458
          BC042190.1 - 18 2528 4188303

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          negative regulation of cell proliferation
          response to muscle activity
          cellular homeostasis
          collagen metabolic process
          multicellular organism growth
          intracellular signal transduction
          multicellular organismal homeostasis
          phosphoprotein phosphatase activity
          receptor binding
          protein tyrosine/serine/threonine phosphatase activity
          protein tyrosine/threonine phosphatase activity
          JUN kinase phosphatase activity
          phosphohistidine phosphatase activity
          inositol bisphosphate phosphatase activity
          hydrolase activity
          MAP kinase tyrosine/serine/threonine phosphatase activity
          inositol-1,3,4-trisphosphate 4-phosphatase activity
          NADP phosphatase activity
          transmembrane receptor protein phosphatase activity
          kinase binding
          protein kinase binding
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
          inositol-4,5-bisphosphate 5-phosphatase activity
          5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
          phosphatidylinositol-4-phosphate phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
          metal ion binding
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
          inositol phosphate phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
          inositol-3,4-bisphosphate 4-phosphatase activity
          inositol-1,3,4-trisphosphate 1-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
          inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
          phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
          IDP phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0