Search
Search

Snx13| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014973.2 | NP_001014973.2 | 26 | 2869 | ||
| GenBank mRNA | AK122352.1 | - | 15 | 1518 | ||
| AK133353.1 | - | 23 | 2409 | |||
| AK164161.1 | - | 26 | 2794 | |||
| BC056394.1 | - | 26 | 2841 | 6852346 | ||