Search
Search

Snx13| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014973.2 | NP_001014973.2 | 3, 4 | 439 | ||
| GenBank mRNA | AK148157.1 | - | 3, 4 | 412 | ||
| AK154425.1 | - | 3, 4 | 441 | |||
| AK164161.1 | - | 3, 4 | 371 | |||
| AK170737.1 | - | 3, 4 | 412 | |||
| BC056394.1 | - | 3, 4 | 411 | 6852346 | ||
| BC067201.1 | - | 3, 4 | 428 | 6825168 | ||
| BY749932.1 | - | 3, 4 | 441 | |||