Search
Search

Tmem260| Gene Symbol: Tmem260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172600.3 | NP_766188.3 | 5 | 672 | ||
| GenBank mRNA | AK031240.1 | - | 6 | 858 | ||
| AK032809.1 | - | 5 | 643 | |||
| AK038175.1 | - | 5 | 628 | |||
| AK040456.1 | - | 4 | 453 | |||
| AK054494.1 | - | 5 | 645 | |||
| AK200005.1 | - | 2 | 77 | |||
| BC059217.1 | - | 5 | 448 | 6408599 | ||