Search
Search

Tmem55b| Gene Symbol: Tmem55b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033271.4 | NP_001028443.1 | 7 | 884 | ||
| GenBank mRNA | AK144251.1 | - | 7 | 884 | ||
| AK152640.1 | - | 7 | 869 | |||
| AK152658.1 | - | 7 | 869 | |||
| AK169026.1 | - | 3 | 2511 | |||
| AK185482.1 | - | 3 | 251 | |||
| BC058171.1 | - | 7 | 833 | |||