Search
Search

Pyroxd1| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183165.3 | NP_898988.2 | 6 | 539 | ||
| GenBank mRNA | AK148411.1 | - | 578 | |||
| AK165677.1 | - | 6 | 542 | |||
| BC022914.1 | - | 6 | 558 | |||
| BC027061.1 | - | 6 | 564 | 5355217 | ||