Search
Search

Amn1| Gene Symbol: Amn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113424.1 | NP_001106895.1 | 7 | 826 | ||
| GenBank mRNA | AK038995.1 | - | 5 | 872 | ||
| AK050169.1 | - | 7 | 1167 | |||
| AK053764.1 | - | 4 | 2761 | |||
| BC026924.1 | - | 5 | 2009 | |||