Search
Search

Plxnb1| Gene Symbol: Plxnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172775.2 | NP_766363.2 | 24 | 4790 | ||
| GenBank mRNA | AB072381.1 | - | 24 | 4634 | ||
| AK129133.1 | - | 13 | 1787 | |||
| BC150701.1 | - | 23 | 4582 | 9087612 | ||