Search
Search

Btnl9| Gene Symbol: Btnl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172793.2 | NP_766381.1 | 10 | 1674 | ||
| GenBank mRNA | AK080918.1 | - | 10 | 1658 | ||
| AK084528.1 | - | 10 | 1675 | |||
| AY177687.1 | - | 10 | 1896 | |||
| BC150808.1 | - | 10 | 1631 | 9087719 | ||
| BC151155.1 | - | 10 | 1631 | 9088043 | ||