Search
Search

Abca12| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 22 | 3464 | ||
| GenBank mRNA | AK076440.1 | - | 3 | 515 | ||
| BC158053.1 | - | 22 | 3277 | 9088162 | ||
| BC158063.1 | - | 22 | 3277 | 9007362 | ||