Search
Search

Map7d1| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145970.1 | NP_001139442.1 | 9 | 1722 | ||
| NM_144941.3 | NP_659190.3 | 9 | 1938 | |||
| GenBank mRNA | AK122462.1 | - | 3 | 3361 | ||
| AK151613.1 | - | 7 | 1453 | |||
| BC019977.1 | - | 4 | 914 | 4187506 | ||
| BC023677.1 | - | 9 | 1913 | 5354497 | ||
| BC023774.1 | - | 9 | 1719 | |||
| CK617787.1 | - | 2 | 393 | |||