Search
Search

Islr| Gene Symbol: Islr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195431.1 | NP_001182360.1 | 2 | 1287 | ||
| NM_012043.4 | NP_036173.1 | 2 | 1419 | |||
| GenBank mRNA | AB024538.1 | - | 2 | 1240 | ||
| BC006602.1 | - | 2 | 1340 | 3966024 | ||