Search
Search

Agpat3| Gene Symbol: Agpat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053014.3 | NP_443747.2 | 7 | 880 | ||
| GenBank mRNA | AB377215.1 | - | 5 | 696 | ||
| AK008965.1 | - | 7 | 1036 | |||
| AK015906.1 | - | 6 | 808 | |||
| AK030607.1 | - | 5 | 672 | |||
| AK075715.1 | - | 7 | 880 | |||
| AK076414.1 | - | 6 | 849 | |||
| AK159110.1 | - | 6 | 1120 | |||
| AY167588.1 | - | 5 | 718 | |||
| BC033444.1 | - | 5 | 558 | |||
| BC052382.1 | - | 6 | 771 | 5703571 | ||
| BC058519.1 | - | 6 | 770 | 5703571 | ||