Search
Search

Cnot11| Gene Symbol: Cnot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028043.2 | NP_082319.1 | 5 | 1159 | ||
| GenBank mRNA | AK010503.1 | - | 5 | 1171 | ||
| AK041342.1 | - | 3 | 484 | |||
| AK144692.1 | - | 5 | 1174 | |||
| AK162118.1 | - | 1465 | ||||
| AK167440.1 | - | 5 | 1150 | |||
| BC049091.1 | - | 5 | 1159 | 6401756 | ||
| BC050812.1 | - | 5 | 1139 | 6773595 | ||