Search
Search

Ebag9| Gene Symbol: Ebag9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019480.4 | NP_062353.3 | 2 | 330 | ||
| GenBank mRNA | AF076524.1 | - | 1 | 13 | ||
| AK004366.1 | - | 1 | 38 | |||
| AK159499.1 | - | 2 | 380 | |||
| AK160149.1 | - | 2 | 499 | |||
| AY009091.1 | - | 2 | 334 | |||
| BC026717.1 | - | 2 | 85 | |||
| BC108397.1 | - | 2 | 330 | 6329278 | ||
| BC119118.1 | - | 2 | 246 | 8733867 | ||
| BC119120.1 | - | 2 | 246 | 8733869 | ||
| BC144740.1 | - | 2 | 280 | 9053288 | ||