Search
Search

Slc15a1| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053079.2 | NP_444309.2 | 10, 11 | 850 | ||
| GenBank mRNA | AF205540.1 | - | 10, 11 | 849 | ||
| AK085471.1 | - | 8, 9 | 1957 | |||
| AK136428.1 | - | 10, 11 | 851 | |||
| BC116248.1 | - | 10, 11 | 840 | 40093541 | ||
| BC116249.1 | - | 10, 11 | 840 | 40093542 | ||