Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RSAD2 SIRNA Assays ›
          Gene Symbol:
          Rsad2
          Gene Name:
          radical S-adenosyl methionine domain containing 2
          Gene Aliases:
          2510004L01Rik, Vig1, cig5
          Chromosome Location:
          Chr.12: 26442743 - 26456478 on Build GRCm38
          UniGene ID:
          Mm.24045
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS226679
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rsad2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021384.4 NP_067359.2 1 260
          GenBank mRNA AF236064.2 - 1 236
          AF442152.1 - 1 221
          AK010906.1 - 1 240
          AK036373.1 - 1 240
          AK149900.1 - 1 242
          AK150069.1 - 1 242
          AK150347.1 - 1 242
          AK150406.1 - 1 240
          AK150425.1 - 1 242
          AK150525.1 - 1 241
          AK150536.1 - 1 242
          AK150616.1 - 1 240
          AK150684.1 - 1 242
          AK150731.1 - 1 242
          AK150767.1 - 1 242
          AK150783.1 - 1 242
          AK150788.1 - 1 242
          AK150812.1 - 1 242
          AK150835.1 - 1 241
          AK151140.1 - 1 242
          AK151185.1 - 1 242
          AK151191.1 - 1 242
          AK151206.1 - 1 242
          AK151286.1 - 1 240
          AK151308.1 - 1 242
          AK151331.1 - 1 242
          AK151339.1 - 1 240
          AK151389.1 - 1 242
          AK151410.1 - 1 241
          AK151412.1 - 1 240
          AK151414.1 - 1 241
          AK151476.1 - 1 242
          AK151502.1 - 1 242
          AK151538.1 - 1 242
          AK151767.1 - 1 242
          AK151770.1 - 1 240
          AK151825.1 - 1 240
          AK151833.1 - 1 240
          AK151846.1 - 1 240
          AK151906.1 - 1 242
          AK151909.1 - 1 240
          AK151940.1 - 1 240
          AK151954.1 - 1 242
          AK151971.1 - 1 242
          AK152050.1 - 1 240
          AK152065.1 - 1 242
          AK152075.1 - 1 242
          AK152235.1 - 1 240
          AK152239.1 - 1 242
          AK152241.1 - 1 242
          AK152310.1 - 1 240
          AK152354.1 - 1 240
          AK152493.1 - 1 242
          AK152600.1 - 1 240
          AK152601.1 - 1 242
          AK152631.1 - 1 240
          AK152644.1 - 1 242
          AK152664.1 - 1 242
          AK152701.1 - 1 242
          AK152796.1 - 1 240
          AK152861.1 - 1 240
          AK152880.1 - 1 240
          AK152895.1 - 1 242
          AK152934.1 - 1 242
          AK152962.1 - 1 240
          AK153003.1 - 1 242
          AK153024.1 - 1 242
          AK153062.1 - 1 244
          AK153077.1 - 1 240
          AK153078.1 - 1 240
          AK153232.1 - 1 242
          AK153234.1 - 1 242
          AK153306.1 - 1 240
          AK153324.1 - 1 242
          AK153350.1 - 1 242
          AK153435.1 - 1 118
          AK153453.1 - 1 241
          AK153556.1 - 1 242
          AK159147.1 - 1 240
          BC057868.1 - 1 233 5327504
          CK623585.1 - 1 260
          CT010378.1 - 1 227

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          immune system process
          response to virus
          regulation of ossification
          positive regulation of toll-like receptor 7 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          CD4-positive, alpha-beta T cell activation
          CD4-positive, alpha-beta T cell differentiation
          negative regulation of viral genome replication
          innate immune response
          negative regulation of protein secretion
          defense response to virus
          positive regulation of T-helper 2 cell cytokine production
          catalytic activity
          protein binding
          protein self-association
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0