Search
Search

Frmd8| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026169.4 | NP_080445.1 | 9 | 1165 | ||
| GenBank mRNA | AK004603.2 | - | 9 | 1188 | ||
| AK011659.1 | - | 4 | 505 | |||
| AK049224.1 | - | 9 | 1180 | |||
| AK049233.1 | - | 9 | 1180 | |||
| AK083902.1 | - | 9 | 1167 | |||
| AK088639.1 | - | 9 | 1177 | |||
| AK148434.1 | - | 1177 | ||||
| AK168669.1 | - | 9 | 1176 | |||
| BC004765.1 | - | 9 | 1233 | 3156148 | ||