Search
Search

Fam45a| Gene Symbol: Fam45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167829.1 | NP_001161301.1 | 3 | 324 | ||
| NM_026437.3 | NP_080713.2 | 3 | 324 | |||
| GenBank mRNA | AK007867.1 | - | 3 | 284 | ||
| AK145769.1 | - | 3 | 290 | |||
| AK171767.1 | - | 3 | 3004 | |||
| BC013514.1 | - | 3 | 283 | 4235293 | ||