Search
Search

Ppap2b| Gene Symbol: Ppap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080555.2 | NP_542122.1 | 2 | 719 | ||
| GenBank mRNA | AK011276.1 | - | 2 | 718 | ||
| AK076183.1 | - | 2 | 415 | |||
| AK159136.1 | - | 2 | 805 | |||
| AK160056.1 | - | 2 | 619 | |||
| BC005558.1 | - | 2 | 706 | 3964817 | ||