Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKR1C21 SIRNA Assays ›
          Gene Symbol:
          Akr1c21
          Gene Name:
          aldo-keto reductase family 1, member C21
          Gene Aliases:
          9430025F20Rik, AI315367
          Chromosome Location:
          Chr.13: 4574075 - 4586543 on Build GRCm38
          UniGene ID:
          Mm.27085
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS233937
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Akr1c21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_029901.2 NP_084177.2 3 373
          GenBank mRNA AI882115.1 - 3 371
          AK020439.1 - 3 373
          AY742217.1 - 3 299
          BC061057.1 - 3 369 30305353
          BY721648.1 - 3 373

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          steroid biosynthetic process
          steroid metabolic process
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          alditol:NADP+ 1-oxidoreductase activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          phenanthrene 9,10-monooxygenase activity
          acetoin dehydrogenase activity
          carboxylic acid binding
          bile acid binding
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          androsterone dehydrogenase activity
          androsterone dehydrogenase (B-specific) activity
          ketosteroid monooxygenase activity
          trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity
          3-ketoglucose-reductase activity
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-blthd:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline