Search
Search

Cetn3| Gene Symbol: Cetn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007684.3 | NP_031710.1 | 4, 5 | 508 | ||
| GenBank mRNA | AK003473.1 | - | 4, 5 | 524 | ||
| AK004532.1 | - | 4, 5 | 520 | |||
| AK006289.1 | - | 4, 5 | 523 | |||
| AK006891.1 | - | 4, 5 | 508 | |||
| AK017505.1 | - | 4, 5 | 548 | |||
| BC002162.1 | - | 4, 5 | 542 | 3485864 | ||
| BC054097.1 | - | 4, 5 | 544 | 30019209 | ||
| Y12474.1 | - | 4, 5 | 459 | |||