Search
Search

Cetn3| Gene Symbol: Cetn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007684.3 | NP_031710.1 | 4 | 356 | ||
| GenBank mRNA | AK003473.1 | - | 4 | 372 | ||
| AK004532.1 | - | 4 | 368 | |||
| AK006289.1 | - | 4 | 371 | |||
| AK006891.1 | - | 4 | 356 | |||
| AK011821.1 | - | 4 | 349 | |||
| AK017505.1 | - | 4 | 396 | |||
| BC002162.1 | - | 4 | 390 | 3485864 | ||
| BC054097.1 | - | 4 | 392 | 30019209 | ||
| Y12474.1 | - | 4 | 307 | |||