Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GREM1 SIRNA Assays ›
          Gene Symbol:
          Grem1
          Gene Name:
          gremlin 1
          Gene Aliases:
          Cktsf1b1, Drm, Grem, ld
          Chromosome Location:
          Chr.2: 113748674 - 113758788 on Build GRCm38
          UniGene ID:
          Mm.166318
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS239454
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Grem1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011824.4 NP_035954.1 2 183
          GenBank mRNA AF045801.1 - 1 24
          AF108189.1 - 1 93
          AK135022.1 - 2 218
          AK157793.1 - 2 184
          AK164871.1 - 2 184
          BC015293.1 - 2 153 4216669
          BY151119.1 - 2 181

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          angiogenesis
          branching involved in ureteric bud morphogenesis
          cell migration involved in sprouting angiogenesis
          positive regulation of receptor internalization
          negative regulation of leukocyte chemotaxis
          positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          signal transduction
          activation of transmembrane receptor protein tyrosine kinase activity
          cell-cell signaling
          positive regulation of cell proliferation
          organ morphogenesis
          proximal/distal pattern formation
          regulation of epithelial to mesenchymal transition
          collagen fibril organization
          negative regulation of cell growth
          embryonic limb morphogenesis
          negative regulation of bone mineralization
          negative regulation of BMP signaling pathway
          negative regulation of chondrocyte differentiation
          regulation of stress-activated MAPK cascade
          negative regulation of osteoblast proliferation
          positive regulation of NF-kappaB import into nucleus
          negative regulation of apoptotic process
          endothelial cell migration
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of bone remodeling
          determination of dorsal identity
          positive regulation of NF-kappaB transcription factor activity
          regulation of focal adhesion assembly
          positive regulation of telomerase activity
          limb development
          negative regulation of pathway-restricted SMAD protein phosphorylation
          ureteric bud formation
          positive regulation of protein tyrosine kinase activity
          signal transduction by p53 class mediator
          negative regulation of monocyte chemotaxis
          negative regulation of canonical Wnt signaling pathway
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of branching involved in ureteric bud morphogenesis
          negative regulation of osteoclast proliferation
          positive regulation of peptidyl-tyrosine autophosphorylation
          negative regulation of bone trabecula formation
          negative regulation of bone mineralization involved in bone maturation
          positive regulation of receptor activity
          positive regulation of cardiac muscle cell differentiation
          cytokine activity
          transmembrane receptor protein tyrosine kinase activator activity
          vascular endothelial growth factor receptor 2 binding
          heparan sulfate proteoglycan binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0