Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERP1 SIRNA Assays ›
          Gene Symbol:
          Serp1
          Gene Name:
          stress-associated endoplasmic reticulum protein 1
          Gene Aliases:
          D3Ucla1, Ramp4
          Chromosome Location:
          Chr.3: 58521969 - 58525884 on Build GRCm38
          UniGene ID:
          Mm.29702
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS241324
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Serp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030685.3 NP_109610.1 1 429
          GenBank mRNA AB041655.1 - 1 298
          AK002653.1 - 1 301
          AK032680.1 - 1 299
          AK050508.1 - 1 269
          AK075776.1 - 1 295
          AK144732.1 - 1 294
          AK148485.1 - 1 300
          AK149786.1 - 1 299
          AK150005.1 - 1 299
          AK150025.1 - 1 299
          AK150368.1 - 1 298
          AK150428.1 - 1 299
          AK150659.1 - 1 294
          AK150714.1 - 1 298
          AK150770.1 - 1 298
          AK150809.1 - 1 268
          AK151323.1 - 1 268
          AK151384.1 - 1 281
          AK151456.1 - 1 299
          AK151669.1 - 1 294
          AK151853.1 - 1 298
          AK152373.1 - 1 294
          AK152383.1 - 1 298
          AK152583.1 - 1 294
          AK152636.1 - 1 299
          AK152806.1 - 1 299
          AK152882.1 - 1 294
          AK152996.1 - 1 299
          AK153056.1 - 1 268
          AK153118.1 - 1 268
          AK153138.1 - 1 294
          AK153286.1 - 1 268
          AK153415.1 - 1 298
          AK159271.1 - 1 299
          AK161595.1 - 1 299
          AK166004.1 - 1 294
          AK166052.1 - 1 274
          AK167482.1 - 1 269
          AK167952.1 - 1 299
          AK168046.1 - 1 299
          AK168569.1 - 1 298
          AK168734.1 - 1 298
          AK168754.1 - 1 298
          AK168947.1 - 1 298
          AK169049.1 - 1 294
          AK169175.1 - 1 299
          AK170734.1 - 1 299
          BC002114.1 - 1 275 3489738
          CV676503.1 - 1 438

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_014445, X75694, BC108314, AB022427, AK125413, and others.
          This siRNA also targets the following transcripts in Rat: NM_030835, BC061854, AB018546, AF100470, AJ238236.

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s)

          skeletal system development
          glucose metabolic process
          protein glycosylation
          transport
          post-embryonic development
          multicellular organism aging
          protein transport
          endoplasmic reticulum unfolded protein response
          positive regulation of insulin secretion
          positive regulation of translation
          positive regulation of organ growth
          muscle organ morphogenesis
          positive regulation of growth hormone secretion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0