Search
Search

Nell1| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037906.2 | NP_001032995.1 | 12 | 1235 | ||
| GenBank mRNA | AK032845.1 | - | 12 | 1517 | ||
| AK046127.1 | - | 564 | ||||
| AY622226.1 | - | 12 | 1235 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK035921.1 | - | 1312 | Chr. - - [-] on Build | ||