Search
Search

Ccdc103| Gene Symbol: Ccdc103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028492.2 | NP_082768.1 | 3 | 175 | ||
| GenBank mRNA | AK006639.1 | - | 3 | 127 | ||
| AK032002.1 | - | 3 | 176 | |||
| BC119102.1 | - | 3 | 174 | 8733851 | ||
| BC119104.1 | - | 3 | 174 | 8733853 | ||
| BC144894.1 | - | 3 | 174 | |||